Gene Transcripts

Gene Symbol
WDR45
Entrez Gene ID
11152
Gene Name
WD repeat domain 45
Gene Aliases
JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
Chromosome Location
Chr. X: 49074433 - 49101121 on Build GRCh38
UniGene ID
Hs.632807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029896.1 NP_001025067.1 9 939
NM_007075.3 NP_009006.2 10 1209
GenBank mRNA AJ005897.1 - 9 895
AK023344.1 - 7 1917
AK225122.1 - 9 918
AK315717.1 - 9 926
AY691428.1 - 8 767
BC000464.2 - 9 933 2822742
BC003037.1 - 9 933 2822742
BC009027.1 - 6 596
BC069206.1 - 10 1209 5801768
CR456725.1 - 8 767

View Full Product Details