Gene Transcripts

Gene Symbol
Tmem52b
Entrez Gene ID
689963
Gene Name
transmembrane protein 52B
Gene Aliases
Chromosome Location
Chr. 4: 163267409 - 163273527 on Build Rnor_6.0
UniGene ID
Rn.155690
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109560.1 NP_001103030.1 2 605
XM_006237102.2 XP_006237164.1 3 542

View Full Product Details