Gene Transcripts
- Gene Symbol
- Kremen1
- Entrez Gene ID
- 114107
- Gene Name
- kringle containing transmembrane protein 1
- Gene Aliases
- Kremen
- Chromosome Location
- Chr. 14: 85441209 - 85503661 on Build Rnor_6.0
- UniGene ID
- Rn.90021
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053649.1 |
NP_446101.1
|
2, 3 |
264 |
|
GenBank mRNA |
AB065090.1 |
- |
2, 3 |
264 |
|
View Full Product Details