Gene Transcripts

Gene Symbol
Snrpd1
Entrez Gene ID
291794
Gene Name
small nuclear ribonucleoprotein D1
Gene Aliases
Chromosome Location
Chr. 18: 1866065 - 1876613 on Build Rnor_6.0
UniGene ID
Rn.18606
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106163.1 NP_001099633.1 4 622
XM_008771943.1 XP_008770165.1 4 514
GenBank mRNA BC167095.1 - 4 515

View Full Product Details