Gene Transcripts

Gene Symbol
NISCH
Entrez Gene ID
11188
Gene Name
nischarin
Gene Aliases
I-1, IR1, IRAS, hIRAS
Chromosome Location
Chr. 3: 52455508 - 52493072 on Build GRCh38
UniGene ID
Hs.435290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276293.1 NP_001263222.1 13 1651
NM_001276294.1 NP_001263223.1 13 1651
NM_007184.3 NP_009115.2 13 1651
XM_005264839.2 XP_005264896.1 12 1500
XM_017005643.1 XP_016861132.1 4 492
GenBank mRNA AB023192.1 - 14 1589
AF058290.1 - 1 114
AF082516.1 - 13 1543
AK001505.1 - 10 1764
AK027428.1 - 13 1529
AK291398.1 - 13 1555
AK302441.1 - 4 539
AK315001.1 - 13 1555
BC038102.2 - 13 1555 5268247
BC054494.1 - 13 1549 6195532
BC056900.1 - 13 1521 6143316
BU177208.1 - 4 402

View Full Product Details