Gene Transcripts
- Gene Symbol
- Cdh15
- Entrez Gene ID
- 361432
- Gene Name
- cadherin 15
- Gene Aliases
- Chromosome Location
- Chr. 19: 55669661 - 55689986 on Build Rnor_6.0
- UniGene ID
- Rn.18406
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207613.1 |
NP_997496.1
|
10 |
1425 |
|
GenBank mRNA |
AB176538.1 |
- |
10 |
1425 |
|
View Full Product Details