Gene Transcripts

Gene Symbol
LOC686142
Entrez Gene ID
686142
Gene Name
similar to Ral guanine nucleotide dissociation stimulator (RalGEF) (RalGDS)
Gene Aliases
RGD1564791
Chromosome Location
Chr. 17: 59697180 - 59704677 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751717.3 XP_003751765.1 3 525
XM_006254133.2 XP_006254195.1 2 1128
XM_006254135.2 XP_006254197.1 2 269

View Full Product Details