Gene Transcripts

Gene Symbol
FMNL1
Entrez Gene ID
752
Gene Name
formin like 1
Gene Aliases
C17orf1, C17orf1B, FHOD4, FMNL, KW-13
Chromosome Location
Chr. 17: 45221728 - 45247322 on Build GRCh38
UniGene ID
Hs.100217
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005892.3 NP_005883.2 23 3173
XM_006722062.3 XP_006722125.1 24 3388
XM_006722063.3 XP_006722126.1 23 3368
XM_006722064.3 XP_006722127.1 24 3358
XM_006722065.3 XP_006722128.1 23 3338
XM_006722066.3 XP_006722129.1 24 3384
XM_006722070.3 XP_006722133.1 23 3364
XM_011525179.2 XP_011523481.1 24 3388
XM_011525180.2 XP_011523482.1 24 3080
XM_011525182.2 XP_011523484.1 24 3385
GenBank mRNA AF432213.1 - 20 2647
AJ008112.1 - 8 1089
AJ008128.1 - 1 29
AK131091.1 - 14 3860
AY278319.1 - 23 3173
BC001710.1 - 8 1077
BC009000.2 - 3 354
BC021906.1 - 11 1681
BC073988.1 - 11 1721
BC111564.1 - 16 1999 40008718
CR456759.1 - 8 1050
FJ534522.1 - 7 972

View Full Product Details