Gene Transcripts

Gene Symbol
KLHL2
Entrez Gene ID
11275
Gene Name
kelch like family member 2
Gene Aliases
ABP-KELCH, MAV, MAYVEN
Chromosome Location
Chr. 4: 165207618 - 165323156 on Build GRCh38
UniGene ID
Hs.388668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161521.1 NP_001154993.1 7 867
NM_001161522.1 NP_001154994.1 6 760
NM_007246.3 NP_009177.3 7 990
XM_011531572.2 XP_011529874.1 8 980
XM_011531575.2 XP_011529877.1 6 812
XM_011531576.2 XP_011529878.1 5 480
XM_017007674.1 XP_016863163.1 8 940
XM_017007675.1 XP_016863164.1 6 705
XM_017007676.1 XP_016863165.1 6 813
XM_017007677.1 XP_016863166.1 5 687
GenBank mRNA AB209727.1 - 3 334
AF059569.1 - 7 839
AK025289.1 - 4 456
AK294103.1 - 7 861
AK294336.1 - 3 289
AK308940.1 - 3 290
AK315372.1 - 7 835
BC022503.1 - 7 972 4791972
BC036468.2 - 7 931 4799622
BX647776.1 - 6 744

View Full Product Details