Gene Transcripts
- Gene Symbol
- TMC6
- Entrez Gene ID
- 11322
- Gene Name
- transmembrane channel like 6
- Gene Aliases
- EV1, EVER1, EVIN1, LAK-4P
- Chromosome Location
- Chr. 17: 78108775 - 78132427 on Build GRCh38
- UniGene ID
- Hs.632227
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127198.2 |
NP_001120670.1
|
13, 14 |
1967 |
|
NM_001321185.1 |
NP_001308114.1
|
13, 14 |
1920 |
|
NM_007267.7 |
NP_009198.4
|
13, 14 |
1871 |
|
XM_005256995.1 |
XP_005257052.1
|
13, 14 |
1996 |
|
XM_011524255.1 |
XP_011522557.1
|
12, 13 |
1898 |
|
XM_011524256.1 |
XP_011522558.1
|
12, 13 |
1772 |
|
XM_011524257.2 |
XP_011522559.1
|
7, 8 |
1016 |
|
XM_017024109.1 |
XP_016879598.1
|
11, 12 |
1687 |
|
GenBank mRNA |
AB002405.2 |
- |
5, 6 |
679 |
|
AK074065.1 |
- |
13, 14 |
1870 |
|
AK310734.1 |
- |
12, 13 |
2291 |
|
AY057379.1 |
- |
12, 13 |
1710 |
|
AY099356.1 |
- |
14, 15 |
1869 |
|
AY236496.1 |
- |
12, 13 |
1836 |
|
BC018346.1 |
- |
6, 7 |
835 |
|
BC023597.2 |
- |
13, 14 |
1851 |
4566166 |
BC035648.1 |
- |
6, 7 |
891 |
5575949 |
View Full Product Details