Gene Transcripts

Gene Symbol
ZNF569
Entrez Gene ID
148266
Gene Name
zinc finger protein 569
Gene Aliases
ZAP1, ZNF
Chromosome Location
Chr. 19: 37411146 - 37469268 on Build GRCh38
UniGene ID
Hs.511848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152484.2 NP_689697.2 6 1747
XM_006723046.2 XP_006723109.1 8 2074
XM_006723047.3 XP_006723110.1 7 1997
XM_006723048.3 XP_006723111.1 7 1921
XM_011526538.2 XP_011524840.1 7 1836
XM_011526539.2 XP_011524841.1 5 1330
XM_017026376.1 XP_016881865.1 6 1582
XM_017026377.1 XP_016881866.1 6 1661
XM_017026378.1 XP_016881867.1 7 1699
XM_017026379.1 XP_016881868.1 4 1548
XM_017026380.1 XP_016881869.1 4 1356
XM_017026381.1 XP_016881870.1 5 1759
XM_017026382.1 XP_016881871.1 4 1456
GenBank mRNA AK056615.1 - 1 544
AK122846.1 - 3 1068
AK289987.1 - 6 1520
AL833408.1 - 6 1748
AY835704.1 - 6 1747
BC059407.1 - 5 1376
BC117219.1 - 2 1004 40125770

View Full Product Details