Gene Transcripts

Gene Symbol
FHL3
Entrez Gene ID
2275
Gene Name
four and a half LIM domains 3
Gene Aliases
SLIM2
Chromosome Location
Chr. 1: 37996765 - 38005515 on Build GRCh38
UniGene ID
Hs.57687
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243878.1 NP_001230807.1 5 1201
NM_004468.4 NP_004459.2 6 1377
XM_017000675.1 XP_016856164.1 3 801
GenBank mRNA AF133732.1 - 5 1198
BC001351.2 - 6 1367 2964682
BC011697.2 - 6 1367 3532981
BC014501.1 - 5 1188 4860329
U60116.2 - 5 1237

View Full Product Details