Gene Transcripts

Gene Symbol
PPA2
Entrez Gene ID
27068
Gene Name
pyrophosphatase (inorganic) 2
Gene Aliases
HSPC124, SID6-306
Chromosome Location
Chr. 4: 105369077 - 105474070 on Build GRCh38
UniGene ID
Hs.654957
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006903.4 NP_008834.3 8, 9 798
NM_176866.2 NP_789842.2 5, 6 579
NM_176867.3 NP_789843.2 3, 4 387
NM_176869.2 NP_789845.1 9, 10 885
GenBank mRNA AB026722.1 - 8, 9 792
AF161473.1 - 10, 11 874
AF217187.1 - 9, 10 879
AK000466.1 - 9, 10 877
AK289607.1 - 8, 9 776
BC022803.1 - 3, 4 363
BC035569.1 - 9, 10 857
BC039462.1 - 5, 6 526
BC057219.1 - 8, 9 791 4289401

View Full Product Details