Gene Transcripts

Gene Symbol
PHF1
Entrez Gene ID
5252
Gene Name
PHD finger protein 1
Gene Aliases
MTF2L2, PCL1, PHF2, TDRD19C, hPHF1
Chromosome Location
Chr. 6: 33410751 - 33416453 on Build GRCh38
UniGene ID
Hs.166204
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002636.4 NP_002627.1 7, 8 925
NM_024165.2 NP_077084.1 7, 8 925
NR_027692.1 7, 8 925
XM_011514662.1 XP_011512964.1 7, 8 842
XM_011514663.1 XP_011512965.1 7, 8 842
XM_011514665.1 XP_011512967.1 7, 8 842
XM_011514666.1 XP_011512968.1 7, 8 842
XM_011514669.1 XP_011512971.1 7, 8 842
XM_011514670.2 XP_011512972.1 7, 8 842
GenBank mRNA AF029678.1 - 7, 8 745
AF052205.1 - 7, 8 741
AK225913.1 - 6, 7 927
AK295412.1 - 7, 8 795
AK299162.1 - 4, 5 424
AK316499.1 - 7, 8 876
BC008834.2 - 7, 8 893 4304525

View Full Product Details