Gene Transcripts

Gene Symbol
CYP26B1
Entrez Gene ID
56603
Gene Name
cytochrome P450 family 26 subfamily B member 1
Gene Aliases
CYP26A2, P450RAI-2, P450RAI2, RHFCA
Chromosome Location
Chr. 2: 72129238 - 72147862 on Build GRCh38
UniGene ID
Hs.91546
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277742.1 NP_001264671.1 4, 5 944
NM_019885.3 NP_063938.1 5, 6 1169
XM_005264433.4 XP_005264490.1 5, 6 1133
XM_011532988.1 XP_011531290.1 4, 5 730
GenBank mRNA AF252297.1 - 5, 6 1141
AK294814.1 - 4, 5 916
AK294933.1 - 4, 5 1139
AK295683.1 - 4, 5 784
AK313433.1 - 5, 6 1144
BC069443.1 - 5, 6 1141 7262360
BC109205.2 - 5, 6 1141 40006245
CR749631.1 - 4, 5 793
FJ467289.1 - 4, 5 916

View Full Product Details