Gene Transcripts

Gene Symbol
PSMB6
Entrez Gene ID
5694
Gene Name
proteasome subunit beta 6
Gene Aliases
DELTA, LMPY, Y
Chromosome Location
Chr. 17: 4796144 - 4798503 on Build GRCh38
UniGene ID
Hs.77060
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270481.1 NP_001257410.1 5 609
NM_002798.2 NP_002789.1 5 609
GenBank mRNA AK311768.1 - 5 572
BC000835.2 - 5 576 3456009
BM553399.1 - 5 556
BQ691763.1 - 5 591
BT009809.1 - 5 558
BU848746.1 - 5 541
CR457065.1 - 5 558
D29012.1 - 5 567
X61971.1 - 3 381

View Full Product Details