Gene Transcripts
- Gene Symbol
- PSMB7
- Entrez Gene ID
- 5695
- Gene Name
- proteasome subunit beta 7
- Gene Aliases
- Z
- Chromosome Location
- Chr. 9: 124353465 - 124415473 on Build GRCh38
- UniGene ID
- Hs.213470
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002799.3 |
NP_002790.1
|
4, 5 |
452 |
|
GenBank mRNA |
AJ420455.1 |
- |
1, 2 |
130 |
|
AK098757.1 |
- |
4, 5 |
421 |
|
AK223007.1 |
- |
4, 5 |
405 |
|
AK223356.1 |
- |
4, 5 |
417 |
|
AK314302.1 |
- |
4, 5 |
421 |
|
BC000509.2 |
- |
4, 5 |
408 |
2822305 |
BC008414.1 |
- |
4, 5 |
418 |
|
BC017116.2 |
- |
3, 4 |
343 |
|
BT007218.1 |
- |
4, 5 |
404 |
|
CR456721.1 |
- |
4, 5 |
404 |
|
D38048.1 |
- |
4, 5 |
418 |
|
GU727627.1 |
- |
4, 5 |
421 |
|
View Full Product Details