Gene Transcripts

Gene Symbol
ATAD3B
Entrez Gene ID
83858
Gene Name
ATPase family, AAA domain containing 3B
Gene Aliases
AAA-TOB3, TOB3
Chromosome Location
Chr. 1: 1471732 - 1509466 on Build GRCh38
UniGene ID
Hs.729021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317238.1 NP_001304167.1 14 1942
NM_031921.5 NP_114127.3 16 1825
XM_011542243.2 XP_011540545.1 15 1561
XM_011542244.1 XP_011540546.1 16 1992
GenBank mRNA AB033099.1 - 14 2031
AK024028.1 - 14 1410
AK127857.1 - 10 2596
AK290586.1 - 16 1817
AL834179.1 - 5 2731
BC002542.2 - 16 1751 3138578
BC018701.1 - 16 1751 3139886

View Full Product Details