Gene Transcripts

Gene Symbol
ICK
Entrez Gene ID
22858
Gene Name
intestinal cell (MAK-like) kinase
Gene Aliases
ECO, LCK2, MRK
Chromosome Location
Chr. 6: 53001299 - 53061864 on Build GRCh38
UniGene ID
Hs.417022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014920.3 NP_055735.1 12 1979
NM_016513.4 NP_057597.2 13 2090
XM_011514419.1 XP_011512721.1 12 2000
XM_011514420.2 XP_011512722.1 13 2142
XM_011514421.1 XP_011512723.1 13 2081
XM_017010485.1 XP_016865974.1 13 2374
XM_017010486.1 XP_016865975.1 14 2253
XM_017010487.1 XP_016865976.1 12 1919
XM_017010488.1 XP_016865977.1 13 2780
XM_017010489.1 XP_016865978.1 14 2295
XM_017010490.1 XP_016865979.1 13 2165
XM_017010491.1 XP_016865980.1 13 2126
XM_017010492.1 XP_016865981.1 12 2170
GenBank mRNA AB023153.1 - 12 1866
AF152469.1 - 12 1947
AF225919.1 - 12 1809
AK074892.1 - 13 2060
BC136420.1 - 11 1640 9020408
BC136421.1 - 11 1640 9020409
BC152464.1 - 12 1866 8862600

View Full Product Details