Gene Transcripts

Gene Symbol
OR5M11
Entrez Gene ID
219487
Gene Name
olfactory receptor family 5 subfamily M member 11
Gene Aliases
OR11-199
Chromosome Location
Chr. 11: 56542340 - 56543257 on Build GRCh38
UniGene ID
Hs.553632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005245.1 NP_001005245.1 1 807
GenBank mRNA BC136951.1 - 1 831
BC136953.1 - 1 831 9020945

View Full Product Details