Gene Transcripts

Gene Symbol
IL36RN
Entrez Gene ID
26525
Gene Name
interleukin 36 receptor antagonist
Gene Aliases
FIL1, FIL1(DELTA), FIL1D, IL-36Ra, IL1F5, IL1HY1, IL1L1, IL1RP3, IL36RA, PSORP, PSORS14
Chromosome Location
Chr. 2: 113058638 - 113064744 on Build GRCh38
UniGene ID
Hs.516301
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012275.2 NP_036407.1 3, 4 271
NM_173170.1 NP_775262.1 3, 4 243
GenBank mRNA AF186094.1 - 3, 4 272
AF201830.1 - 2, 3 137
AF230377.1 - 2, 3 110
AJ242737.1 - 3, 4 191
AJ242738.1 - 3, 4 182
AK290249.1 - 3, 4 181
AY359117.1 - 3, 4 176
BC024747.1 - 3, 4 243 4996939

View Full Product Details