Gene Transcripts

Gene Symbol
NBPF15
Entrez Gene ID
284565
Gene Name
neuroblastoma breakpoint family member 15
Gene Aliases
AB14, AG3, NBPF16
Chromosome Location
Chr. 1: 144421386 - 144459464 on Build GRCh38
UniGene ID
Hs.523572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170755.2 NP_001164226.1 22 3198
NM_173638.4 NP_775909.2 19 2668
XM_017001077.1 XP_016856566.1 17 2316
GenBank mRNA AK290142.1 - 11 1370
AK290302.1 - 16 2127
AK299360.1 - 13 1708
BC023087.1 - 19 2525 3918932

Gene Symbol
LOC102724250
Entrez Gene ID
102724250
Gene Name
neuroblastoma breakpoint family member 1
Gene Aliases
Chromosome Location
Chr. 1|NT_187366: 4611 - 41174 on Build GRCh38
UniGene ID
Hs.650616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011546188.2 XP_011544490.1 22 11771

Gene Symbol
NBPF1
Entrez Gene ID
55672
Gene Name
neuroblastoma breakpoint family member 1
Gene Aliases
AB13, AB14, AB23, AD2, NBG, NBPF
Chromosome Location
Chr. 1: 16562427 - 16613605 on Build GRCh38
UniGene ID
Hs.467587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017940.4 NP_060410.2 4674
GenBank mRNA AB051480.1 - 21 2731
AF379624.1 - 23 2929
BC034418.1 - 16 2236

Gene Symbol
NBPF10
Entrez Gene ID
100132406
Gene Name
neuroblastoma breakpoint family member 10
Gene Aliases
AB6, AG1, NBPF9
Chromosome Location
Chr. 1: 146066318 - 146144804 on Build GRCh38
UniGene ID
Hs.714127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF379616.1 - 9 1437
AF419616.1 - 23 2822
BX538005.1 - 28 4134

Gene Symbol
NBPF14
Entrez Gene ID
25832
Gene Name
neuroblastoma breakpoint family member 14
Gene Aliases
DJ328E19.C1.1, NBPF
Chromosome Location
Chr. 1: 148531385 - 148607636 on Build GRCh38
UniGene ID
Hs.534675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF379626.1 - 12 1533
AK095459.1 - 12 2319

Gene Symbol
NBPF19
Entrez Gene ID
101060226
Gene Name
neuroblastoma breakpoint family member 19
Gene Aliases
Chromosome Location
Chr. 1: 149472955 - 149556256 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017003019.1 XP_016858508.1 17 2312
GenBank mRNA AK292666.1 - 8 1065
AK295639.1 - 23 3250
CR936830.1 - 5 3725

Gene Symbol
NBPF20
Entrez Gene ID
100288142
Gene Name
neuroblastoma breakpoint family member 20
Gene Aliases
Chromosome Location
Chr. 1: 145289900 - 145405778 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278267.1 NP_001265196.1 16847
GenBank mRNA AK299612.1 - 8 1021

Gene Symbol
NBPF9
Entrez Gene ID
400818
Gene Name
neuroblastoma breakpoint family member 9
Gene Aliases
AE01
Chromosome Location
Chr. 1: 149054033 - 149090861 on Build GRCh38
UniGene ID
Hs.445080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037675.3 NP_001032764.2 21 2927
NM_001277444.1 NP_001264373.1 25 3434
XM_006711317.2 XP_006711380.1 26 3662
GenBank mRNA AF419619.1 - 2 290
AK294414.1 - 19 2592
BQ436001.1 - 5 661

View Full Product Details