Gene Transcripts

Gene Symbol
TMEM215
Entrez Gene ID
401498
Gene Name
transmembrane protein 215
Gene Aliases
Chromosome Location
Chr. 9: 32783499 - 32789201 on Build GRCh38
UniGene ID
Hs.522063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212558.2 NP_997723.2 2 920
GenBank mRNA AK125320.1 - 2 920
BX647550.1 - 2 879
CR749589.1 - 1 741

View Full Product Details