Gene Transcripts

Gene Symbol
TARP
Entrez Gene ID
445347
Gene Name
TCR gamma alternate reading frame protein
Gene Aliases
CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
Chromosome Location
Chr. 7: 38239577 - 38276741 on Build GRCh38
UniGene ID
Hs.534032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003799.1 NP_001003799.1 2 151
NM_001003806.1 NP_001003806.1 2 151
XM_011515407.2 XP_011513709.1 3 371
XM_017012226.1 XP_016867715.1 3 306
XM_017012227.1 XP_016867716.1 2 361
XM_017012228.1 XP_016867717.1 4 752
XM_017012229.1 XP_016867718.1 2 573
XM_017012230.1 XP_016867719.1 2 177
GenBank mRNA AF151103.1 - 2 151
BC030554.1 - 4 835
BC105589.1 - 3 369 4272944
BF674593.1 - 2 141
EU395807.1 - 2 130
JQ956403.1 - 3 293
KC170727.1 - 5 508
M16804.1 - 3 430
M17324.1 - 1 90
M17325.1 - 1 70
M27331.1 - 1 91
M27332.1 - 1 91
M27334.1 - 1 91
S73842.1 - 3 331
X06774.1 - 4 611
X06775.1 - 2 181
X06776.1 - 3 366
X72500.1 - 4 517
X74774.1 - 3 617
X74775.1 - 2 451
Y00790.1 - 4 529

Gene Symbol
TRGC2
Entrez Gene ID
6967
Gene Name
T cell receptor gamma constant 2
Gene Aliases
TCRGC2, TRGC2(2X), TRGC2(3X)
Chromosome Location
Chr. 7: 38240024 - 38249572 on Build GRCh38
UniGene ID
Hs.731375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA M13231.1 - 4 500

Gene Symbol
TRGV9
Entrez Gene ID
6983
Gene Name
T cell receptor gamma variable 9
Gene Aliases
TCRGV9, TRGC1, V2
Chromosome Location
Chr. 7: 38317017 - 38317492 on Build GRCh38
UniGene ID
Hs.534032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC062761.1 - 2 146
BC072387.1 - 3 468
BC072396.1 - 5 894
M16768.1 - 4 654

View Full Product Details