Gene Transcripts

Gene Symbol
KIFAP3
Entrez Gene ID
22920
Gene Name
kinesin associated protein 3
Gene Aliases
FLA3, KAP-1, KAP-3, KAP3, SMAP, Smg-GDS, dJ190I16.1
Chromosome Location
Chr. 1: 169921329 - 170074741 on Build GRCh38
UniGene ID
Hs.433442
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204514.1 NP_001191443.1 10 1375
NM_001204516.1 NP_001191445.1 8 1079
NM_001204517.1 NP_001191446.1 9 2320
NM_014970.3 NP_055785.2 9 1211
XM_005244970.1 XP_005245027.1 9 1214
XM_011509307.2 XP_011507609.2 9 1214
GenBank mRNA AK026471.1 - 9 990
AK122897.1 - 9 2322
AK297368.1 - 8 1006
AK301912.1 - 2 178
AK303052.1 - 10 1332
AK315586.1 - 9 1125
BC028679.2 - 9 1168 4829354
BC048012.1 - 8 1123
U59919.1 - 9 1128

View Full Product Details