Gene Transcripts

Gene Symbol
CMTM6
Entrez Gene ID
54918
Gene Name
CKLF like MARVEL transmembrane domain containing 6
Gene Aliases
CKLFSF6, PRO2219
Chromosome Location
Chr. 3: 32481312 - 32502911 on Build GRCh38
UniGene ID
Hs.380627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017801.2 NP_060271.1 2 343
GenBank mRNA AF479261.1 - 2 840
AK000403.1 - 2 284
BC002797.2 - 2 299 3633497
CR457231.1 - 2 177

View Full Product Details