Gene Transcripts

Gene Symbol
SLC50A1
Entrez Gene ID
55974
Gene Name
solute carrier family 50 member 1
Gene Aliases
HsSWEET1, RAG1AP1, SCP, SWEET1, slv
Chromosome Location
Chr. 1: 155135253 - 155138858 on Build GRCh38
UniGene ID
Hs.292154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122839.1 NP_001116311.1 3 343
NM_001287586.1 NP_001274515.1 2 246
NM_001287587.1 NP_001274516.1 3 363
NM_001287591.1 NP_001274520.1 3 418
NM_018845.3 NP_061333.2 3 343
XM_005245339.4 XP_005245396.1 3 529
XM_005245340.3 XP_005245397.1 3 610
XM_005245341.4 XP_005245398.1 3 561
XM_005245342.3 XP_005245399.1 2 169
XM_006711453.2 XP_006711516.1 3 343
GenBank mRNA AF126023.1 - 3 303
AK289551.1 - 3 344
BC005943.1 - 3 287 4076256
BC009621.1 - 3 305 3896154
BE958344.1 - 202
BM806886.1 - 3 405
BP248307.1 - 3 488
BU160680.1 - 3 285
CR457148.1 - 3 243
DA113832.1 - 3 344

View Full Product Details