Gene Transcripts

Gene Symbol
MRPS24
Entrez Gene ID
64951
Gene Name
mitochondrial ribosomal protein S24
Gene Aliases
HSPC335, MRP-S24, S24mt, bMRP-47, bMRP47
Chromosome Location
Chr. 7: 43866558 - 43869546 on Build GRCh38
UniGene ID
Hs.284286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032014.2 NP_114403.1 4 586
GenBank mRNA AF161453.1 - 4 513
BC012167.1 - 4 538 4637888
BC054865.1 - 4 553 6456699

Gene Symbol
URGCP-MRPS24
Entrez Gene ID
100534592
Gene Name
URGCP-MRPS24 readthrough
Gene Aliases
Chromosome Location
Chr. 7: 43866558 - 43906632 on Build GRCh38
UniGene ID
Hs.663312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204871.1 NP_001191800.1 7 728

View Full Product Details