Gene Transcripts

Gene Symbol
SNRPC
Entrez Gene ID
6631
Gene Name
small nuclear ribonucleoprotein polypeptide C
Gene Aliases
U1C, Yhc1
Chromosome Location
Chr. 6: 34757094 - 34773857 on Build GRCh38
UniGene ID
Hs.1063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003093.2 NP_003084.1 6 1057
NR_029472.1 5 1014
GenBank mRNA AK223098.1 - 6 631
BU153661.1 - 5 553
BU508721.1 - 5 583
X12517.1 - 6 622

View Full Product Details