Gene Transcripts
- Gene Symbol
- LOC102724467
- Entrez Gene ID
- 102724467
- Gene Name
- uncharacterized LOC102724467
- Gene Aliases
- Chromosome Location
- Chr. 16: 87766331 - 87779148 on Build GRCh38
- UniGene ID
- Hs.587857
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120309.1 |
|
1 |
72 |
|
GenBank mRNA |
DB094384.1 |
- |
1 |
74 |
|
DB100335.1 |
- |
1 |
298 |
|
View Full Product Details