Gene Transcripts

Gene Symbol
NOL4
Entrez Gene ID
8715
Gene Name
nucleolar protein 4
Gene Aliases
CT125, HRIHFB2255, NOLP
Chromosome Location
Chr. 18: 33851100 - 34224834 on Build GRCh38
UniGene ID
Hs.514795
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006722563.2 XP_006722626.1 1 653
XM_011526237.1 XP_011524539.1 1 653
XM_011526238.1 XP_011524540.1 1 653
XM_017026055.1 XP_016881544.1 1 653
XM_017026059.1 XP_016881548.1 1 652

View Full Product Details