Gene Transcripts

Gene Symbol
STK38L
Entrez Gene ID
23012
Gene Name
serine/threonine kinase 38 like
Gene Aliases
NDR2
Chromosome Location
Chr. 12: 27244143 - 27325959 on Build GRCh38
UniGene ID
Hs.184523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015000.3 NP_055815.1 11, 12 1272
XM_005253342.3 XP_005253399.1 10, 11 1151
XM_006719058.3 XP_006719121.1 12, 13 1253
XM_006719059.3 XP_006719122.1 12, 13 1232
XM_011520613.2 XP_011518915.1 10, 11 966
XM_017019036.1 XP_016874525.1 11, 12 1228
GenBank mRNA AB023182.2 - 11, 12 1357
AK291055.1 - 11, 12 1162
AK303373.1 - 1, 2 432
AK304755.1 - 9, 10 856
AK304862.1 - 5, 6 568
BC028603.1 - 11, 12 1246 4823429
BX390969.1 - 4, 5 357

View Full Product Details