Gene Transcripts

Gene Symbol
MAGIX
Entrez Gene ID
79917
Gene Name
MAGI family member, X-linked
Gene Aliases
JM10, PDZX
Chromosome Location
Chr. X: 49162569 - 49168157 on Build GRCh38
UniGene ID
Hs.193170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099681.2 NP_001093151.2 5 1430
NM_001099682.2 NP_001093152.2 5 1415
NM_024859.3 NP_079135.3 6 1658
GenBank mRNA AK025340.1 - 4 1513

View Full Product Details