Gene Transcripts

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
Chromosome Location
Chr. 12: 110372900 - 110403730 on Build GRCh38
UniGene ID
Hs.719935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137664.1 NP_001131136.1 7 991
NM_016238.2 NP_057322.2 7 991
XM_005253903.3 XP_005253960.1 6 857
XM_011538454.2 XP_011536756.1 7 1219
XM_017019425.1 XP_016874914.1 7 991
XM_017019426.1 XP_016874915.1 7 986
XM_017019427.1 XP_016874916.1 7 1785
XM_017019428.1 XP_016874917.1 6 874
GenBank mRNA AF191340.1 - 7 888
AK021809.1 - 7 970
AL137586.1 - 1 45
BC002941.2 - 2 135
BC009498.2 - 5 561
BC017316.2 - 3 337
BC040130.1 - 7 906
BC098264.1 - 7 887
BC098295.1 - 7 887
BC098362.1 - 7 887
BC099732.1 - 7 887
BC111798.1 - 7 887
BC148236.2 - 7 888 40024981

View Full Product Details