Gene Transcripts

Gene Symbol
SYCE2
Entrez Gene ID
256126
Gene Name
synaptonemal complex central element protein 2
Gene Aliases
CESC1
Chromosome Location
Chr. 19: 12898229 - 12919674 on Build GRCh38
UniGene ID
Hs.655173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105578.1 NP_001099048.1 3 249
XM_005259848.4 XP_005259905.1 3 321
XM_011527882.2 XP_011526184.1 3 651
XM_011527883.2 XP_011526185.1 3 651
GenBank mRNA AK097443.1 - 2 1502
AK302373.1 - 3 239
BC035819.2 - 2 218
DB449708.1 - 3 253

View Full Product Details