Gene Transcripts

Gene Symbol
MRPS9
Entrez Gene ID
64965
Gene Name
mitochondrial ribosomal protein S9
Gene Aliases
MRP-S9, RPMS9, S9mt
Chromosome Location
Chr. 2: 105038025 - 105099960 on Build GRCh38
UniGene ID
Hs.590900
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182640.2 NP_872578.1 2 219
GenBank mRNA AK001451.1 - 2 161
AK130078.1 - 2 160
AV729381.1 - 2 196
BC036575.1 - 2 172
BC057240.1 - 2 173 6495350
DT217261.1 - 2 219

View Full Product Details