Gene Transcripts
- Gene Symbol
- SPRR2D
- Entrez Gene ID
- 6703
- Gene Name
- small proline rich protein 2D
- Gene Aliases
- Chromosome Location
- Chr. 1: 153039725 - 153041144 on Build GRCh38
- UniGene ID
- Hs.505327
- Species
- Homo sapiens
- Gene Symbol
- SPRR2A
- Entrez Gene ID
- 6700
- Gene Name
- small proline rich protein 2A
- Gene Aliases
- Chromosome Location
- Chr. 1: 153056120 - 153057512 on Build GRCh38
- UniGene ID
- Hs.355542
- Species
- Homo sapiens
- Gene Symbol
- SPRR2B
- Entrez Gene ID
- 6701
- Gene Name
- small proline rich protein 2B
- Gene Aliases
- Chromosome Location
- Chr. 1: 153070226 - 153071608 on Build GRCh38
- UniGene ID
- Hs.568239
- Species
- Homo sapiens
- Gene Symbol
- SPRR2C
- Entrez Gene ID
- 6702
- Gene Name
- small proline rich protein 2C (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 1: 153140118 - 153141493 on Build GRCh38
- UniGene ID
- Hs.2421
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003062.1 |
|
2 |
238 |
|
GenBank mRNA |
BC120937.1 |
- |
2 |
196 |
|
M21539.1 |
- |
2 |
238 |
|
View Full Product Details