Gene Transcripts

Gene Symbol
CCZ1
Entrez Gene ID
51622
Gene Name
CCZ1 homolog, vacuolar protein trafficking and biogenesis associated
Gene Aliases
C7orf28A, CCZ1A, CGI-43, H_DJ1163J12.2
Chromosome Location
Chr. 7: 5898710 - 5925974 on Build GRCh38
UniGene ID
Hs.530000
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015622.5 NP_056437.4 8, 9 866
GenBank mRNA AF151801.1 - 8, 9 910
BC132749.1 - 8, 9 851 40146771
BC132751.1 - 8, 9 851 40146773
CN310516.1 - 5, 6 558

Gene Symbol
CCZ1B
Entrez Gene ID
221960
Gene Name
CCZ1 homolog B, vacuolar protein trafficking and biogenesis associated
Gene Aliases
C7orf28B, H_NH0577018.2
Chromosome Location
Chr. 7: 6798935 - 6826295 on Build GRCh38
UniGene ID
Hs.567779
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198097.3 NP_932765.1 8, 9 874
GenBank mRNA AK292326.1 - 8, 9 842
AK296026.1 - 7, 8 680
AL050215.1 - 6, 7 516
BC010130.2 - 8, 9 809 3953739

View Full Product Details