Gene Transcripts

Gene Symbol
LEPROTL1
Entrez Gene ID
23484
Gene Name
leptin receptor overlapping transcript-like 1
Gene Aliases
HSPC112, Vps55, my047
Chromosome Location
Chr. 8: 30095406 - 30137706 on Build GRCh38
UniGene ID
Hs.146585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128208.1 NP_001121680.1 3 243
NM_015344.2 NP_056159.2 3 243
GenBank mRNA AF063605.1 - 3 220
AF161461.1 - 3 177
AK075363.1 - 3 187
AY277595.1 - 3 215
AY358674.1 - 3 215
BC000642.2 - 3 226 3344487
CR541735.1 - 2 136
CR541760.1 - 2 136
DA258969.1 - 3 244

View Full Product Details