Gene Transcripts

Gene Symbol
PRKD3
Entrez Gene ID
23683
Gene Name
protein kinase D3
Gene Aliases
EPK2, PKC-NU, PKD3, PRKCN, nPKC-NU
Chromosome Location
Chr. 2: 37250502 - 37324803 on Build GRCh38
UniGene ID
Hs.660757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005813.4 NP_005804.1 14 2499
XM_005264237.3 XP_005264294.1 15 2772
XM_005264238.3 XP_005264295.1 14 1748
XM_017003741.1 XP_016859230.1 16 2777
XM_017003742.1 XP_016859231.1 15 1816
XM_017003743.1 XP_016859232.1 12 1498
XM_017003744.1 XP_016859233.1 11 1246
GenBank mRNA AB015982.2 - 14 2499
AK027585.1 - 11 1308
AK122883.1 - 7 2173

View Full Product Details