Gene Transcripts

Gene Symbol
DCUN1D4
Entrez Gene ID
23142
Gene Name
defective in cullin neddylation 1 domain containing 4
Gene Aliases
Chromosome Location
Chr. 4: 51843000 - 51916837 on Build GRCh38
UniGene ID
Hs.605388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040402.2 NP_001035492.1 7 688
NM_001287755.1 NP_001274684.1 7 584
NM_001287757.1 NP_001274686.1 8 851
NM_015115.3 NP_055930.2 7 688
XM_005265731.3 XP_005265788.1 7 578
XM_011534379.2 XP_011532681.1 7 1111
XM_011534380.1 XP_011532682.1 7 447
XM_017007912.1 XP_016863401.1 7 541
XM_017007913.1 XP_016863402.1 6 1026
GenBank mRNA AK027051.1 - 7 507
AK124346.1 - 6 1256
AK294894.1 - 7 584
AK294896.1 - 8 698
BC053897.1 - 7 554 6173607
BX537944.1 - 7 515
D87466.1 - 7 498

View Full Product Details