Gene Transcripts

Gene Symbol
Cxcr6
Entrez Gene ID
80901
Gene Name
chemokine (C-X-C motif) receptor 6
Gene Aliases
BB217514, BONZO, STRL33
Chromosome Location
Chr. 9: 123806467 - 123811756 on Build GRCm38
UniGene ID
Mm.124289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030712.4 NP_109637.3 2 983
GenBank mRNA AF301018.1 - 2 986
AF305709.1 - 2 986
AK052901.1 - 2 981
AK137703.1 - 2 962
AK153914.1 - 2 982
JC209414.1 - 2 983

View Full Product Details