Gene Transcripts

Gene Symbol
C2cd4d
Entrez Gene ID
271944
Gene Name
C2 calcium-dependent domain containing 4D
Gene Aliases
Gm659
Chromosome Location
Chr. 3: 94362444 - 94364567 on Build GRCm38
UniGene ID
Mm.271006
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136117.1 NP_001129589.1 2 991

View Full Product Details