Gene Transcripts

Gene Symbol
Cxx1b
Entrez Gene ID
553127
Gene Name
CAAX box 1B
Gene Aliases
Mart8c
Chromosome Location
Chr. X: 53669177 - 53670408 on Build GRCm38
UniGene ID
Mm.328602
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018063.1 NP_001018073.1 1 1181
GenBank mRNA AK003646.1 - 1 1160
BC029838.1 - 1 1181 3967024

View Full Product Details