Gene Transcripts

Gene Symbol
Pip4k2b
Entrez Gene ID
108083
Gene Name
phosphatidylinositol-5-phosphate 4-kinase, type II, beta
Gene Aliases
AI848124, PI5P4Kbeta, Pip5k2b, c11
Chromosome Location
Chr. 11: 97715157 - 97744704 on Build GRCm38
UniGene ID
Mm.39700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054051.1 NP_473392.1 4 588
GenBank mRNA AK089414.1 - 4 578
AK146559.1 - 4 638
AK170392.1 - 4 515
AK171497.1 - 4 578
AY050219.1 - 4 417
BC047282.1 - 4 588 6491155

View Full Product Details