Gene Transcripts

Gene Symbol
Coq10a
Entrez Gene ID
210582
Gene Name
coenzyme Q10 homolog A (yeast)
Gene Aliases
Gm1
Chromosome Location
Chr. 10: 128363096 - 128370037 on Build GRCm38
UniGene ID
Mm.296876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081040.1 NP_001074509.1 3 1619


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CK346052.1 - 261 Chr. - - [-]

View Full Product Details