Gene Transcripts

Gene Symbol
Mia
Entrez Gene ID
12587
Gene Name
melanoma inhibitory activity
Gene Aliases
CD-RAP, Cdrap, Mia1
Chromosome Location
Chr. 7: 27179741 - 27181149 on Build GRCm38
UniGene ID
Mm.262447
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019394.3 NP_062267.2 4 542
GenBank mRNA AK135151.1 - 3 942
BC009815.1 - 3 926
CK794645.1 - 4 536
X94322.1 - 4 546

View Full Product Details