Gene Transcripts

Gene Symbol
Srrm3
Entrez Gene ID
58212
Gene Name
serine/arginine repetitive matrix 3
Gene Aliases
2900083I11Rik, AB041661, SRm300-like, Srrm2l
Chromosome Location
Chr. 5: 135806861 - 135874773 on Build GRCm38
UniGene ID
Mm.271230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021403.3 NP_067378.2 8, 9 1192
GenBank mRNA AK013812.1 - 5, 6 338
BC051947.1 - 8, 9 799 5696610
BC058385.1 - 8, 9 1192

View Full Product Details