Gene Transcripts

Gene Symbol
Sp9
Entrez Gene ID
381373
Gene Name
trans-acting transcription factor 9
Gene Aliases
Chromosome Location
Chr. 2: 73271926 - 73275771 on Build GRCm38
UniGene ID
Mm.330293
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005343.2 NP_001005343.1 2 2137
GenBank mRNA AK135123.1 - 2 2136
AY591908.1 - 2 2149

View Full Product Details