Gene Transcripts

Gene Symbol
Vmn2r99
Entrez Gene ID
665376
Gene Name
vomeronasal 2, receptor 99
Gene Aliases
EG665376
Chromosome Location
Chr. 17: 19362135 - 19394590 on Build GRCm38
UniGene ID
Mm.469859
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104551.2 NP_001098021.2 4 1324

View Full Product Details